Platinum(IV) nitrate-Lösung (15% Pt) (w/w)

Lieferant: Thermo Fisher Scientific
Gefahr

H37737.03
H37737.03EA 75.7 EUR
H37737.03
Platinum(IV) nitrate-Lösung (15% Pt) (w/w)
Platinum(IV) nitrate
Formel: N₄O₁₂Pt
Lagertemperatur: Raumtemperatur
MDL: MFCD25372548
CAS-Nummer: 18496-40-7
EINECS: 432-400-1
UN: 3093
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR