Triphos 97+%
Lieferant: Thermo Scientific
Synonyme:
(2-((Diphenylphosphino)methyl)-2-methylpropane-1,3-diyl)bis(diphenylphosphine), 1,1,1-Tris(diphenylphosphinomethyl)ethane
H37863.06
H37863.06EA
268
EUR
H37863.06
Triphos 97+%
Triphos
Purity: 97+%
CAS No.: 22031-12-5
Molecular Formula: C41H39P3
Formula Weight: 624.67
Physical Form: Powder
Sensitivity: Air Sensitive
Appearance: White
Melting Point: 99-102°
MDL No.: MFCD00014084
Formel:
C₄₁H₃₉P₃ Molecular Weight: 624,684 g/mol Schmelzpunkt: 99…102 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014084 CAS-Nummer: 22031-12-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...