2-Amino-5-brom-3-iodpyridin 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 5-Brom-3-iodpyridin-2-amin, 5-Brom-3-iod-2-pyridylamin

H50002.06
H50002.06EA 392 EUR
H50002.06
2-Amino-5-brom-3-iodpyridin 98%
2-Amino-5-brom-3-iodpyridin

Purity: 98% CAS No.: 381233-96-1 Molecular Formula: C5H4BrIN2 Formula Weight: 298.91 Melting Point: 113-115° MDL No.: MFCD06659000

Formel: C₅H₄BrIN₂
Molecular Weight: 298,91 g/mol
Schmelzpunkt: 113…115 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06659000
CAS-Nummer: 381233-96-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR