3-Amino-4-formylpyridin 95%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
3-Amino-pyridine-4-carbaldehyde, 3-Aminoisonicotinaldehyde, 3-Aminopyridine-4-carboxaldehyde
H50024.03
H50024.03EA
134
EUR
H50024.03
3-Amino-4-formylpyridin 95%
3-Amino-4-formylpyridin
Purity: 95%
CAS No.: 55279-29-3
Molecular Formula: C6H6N2O
Formula Weight: 122.13
Melting Point: ca 92°
MDL No.: MFCD06410684
Formel:
C₆H₆N₂O Molecular Weight: 122.13 g/mol Schmelzpunkt: ∼ 92 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06410684 CAS-Nummer: 55279-29-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...