4-Aminonicotinsäure 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-Aminopyridin-3-carbonsäure, 4-Amino-3-pyridincarbonsäure

H50032.06
H50032.06EA 73.6 EUR
H50032.06
4-Aminonicotinsäure 98%
4-Aminonicotinsäure

Purity: 98% CAS No.: 7418-65-7 Molecular Formula: C6H6N2O2 Formula Weight: 138.13 Melting Point: 300° MDL No.: MFCD00234183

Formel: C₆H₆N₂O₂
Molecular Weight: 138,13 g/mol
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00234183
CAS-Nummer: 7418-65-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR