5-Bromo-1H-pyrrolo[2,3-b]pyridine 96%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
5-Bromo-7-azaindole
H50040.03
H50040.03EA
23.9
EUR
H50040.03
5-Bromo-1H-pyrrolo[2,3-b]pyridine 96%
5-Bromo-1H-pyrrolo[2,3-b]pyridine
Purity: 96%
CAS No.: 183208-35-7
Molecular Formula: C7H5BrN2
Formula Weight: 197.04
Melting Point: 178-179°
MDL No.: MFCD06659677
Formel:
C₇H₅BrN₂ Molecular Weight: 197,04 g/mol Schmelzpunkt: 178…179 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06659677 CAS-Nummer: 183208-35-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...