6-Bromo-2-chloro-1,8-naphthyridine 96%

Lieferant: Thermo Scientific
Achtung

H50043.03
H50043.03EA 138 EUR
H50043.03
6-Bromo-2-chloro-1,8-naphthyridine 96%
6-Bromo-2-chloro-1,8-naphthyridine

Purity: 96% CAS No.: 902837-40-5 Molecular Formula: C8H4BrClN2 Formula Weight: 243.49 MDL No.: MFCD07781152

Formel: C₈H₄BrClN₂
Molecular Weight: 243,49 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07781152
CAS-Nummer: 902837-40-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR