2-Chloro-4-iodopyridine-3-carboxaldehyde 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-Chloro-4-iodonicotinaldehyde
H50048.06
H50048.06EA
328
EUR
H50048.06
2-Chloro-4-iodopyridine-3-carboxaldehyde 98%
2-Chloro-4-iodopyridine-3-carboxaldehyde
Purity: 98%
CAS No.: 153034-90-3
Molecular Formula: C6H3ClINO
Formula Weight: 267.45
MDL No.: MFCD03095294
Formel:
C₆H₃ClINO Molecular Weight: 267,45 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03095294 CAS-Nummer: 153034-90-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...