(2-Chlor-4-iod-3-pyridinyl)methanol 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H50052.03
H50052.03EA
91
EUR
H50052.03
(2-Chlor-4-iod-3-pyridinyl)methanol 97%
(2-Chlor-4-iod-3-pyridinyl)methanol
Purity: 97%
CAS No.: 884494-44-4
Molecular Formula: C6H5ClINO
Formula Weight: 269.47
MDL No.: MFCD03095296
Formel:
C₆H₅ClINO Lagertemperatur: Raumtemperatur |
MDL:
MFCD03095296 CAS-Nummer: 884494-44-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...