2-Methoxy-3-pyridinboronsäurepinakolester ≥97%
Lieferant: Thermo Fisher Scientific
H50060.03
H50060.03EA
131
EUR
H50060.03
2-Methoxy-3-pyridinboronsäurepinakolester ≥97%
2-Methoxy-3-pyridinboronsäurepinakolester
Purity: 97%
CAS No.: 532391-31-4
Molecular Formula: C12H18BNO3
Formula Weight: 235.09
MDL No.: MFCD05663507