2-(Boc-amino)pyridine-5-carboxaldehyde 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
tert-Butyl (5-formylpyridin-2-yl)carbamate, 6-Aminonicotinaldehyde, 6-BOC protected
H50075.03
H50075.03EA
147
EUR
H50075.03
2-(Boc-amino)pyridine-5-carboxaldehyde 97%
2-(Boc-amino)pyridine-5-carboxaldehyde
Purity: 97%
CAS No.: 199296-40-7
Molecular Formula: C11H14N2O3
Formula Weight: 222.25
MDL No.: MFCD08064228
Formel:
C₁₁H₁₄N₂O₃ Molecular Weight: 222.25 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08064228 CAS-Nummer: 199296-40-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...