(2-Chlor-4-pyridinyl)methanol ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: (2-Chloropyridin-4-yl)methanol, 2-Chloro-4-(hydroxymethyl)pyridine, 2-Chloropyridine-4-methanol

H50099.06
H50099.06EA 265 EUR
H50099.06
(2-Chlor-4-pyridinyl)methanol ≥97%
(2-Chlor-4-pyridinyl)methanol

Purity: 97% CAS No.: 100704-10-7 Molecular Formula: C6H6ClNO Formula Weight: 143.57 Melting Point: 64-66° MDL No.: MFCD06858767

Formel: C₆H₆ClNO
Molecular Weight: 143,57 g/mol
Schmelzpunkt: 64…66 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06858767
CAS-Nummer: 100704-10-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR