2-n-Propoxypyridine-3-carboxaldehyde 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-Propoxynicotinaldehyde
H50114.03
H50114.03EA
70
EUR
H50114.03
2-n-Propoxypyridine-3-carboxaldehyde 98%
2-n-Propoxypyridine-3-carboxaldehyde
Purity: 98%
CAS No.: 885278-12-6
Molecular Formula: C9H11NO2
Formula Weight: 165.19
MDL No.: MFCD06804561
Formel:
C₉H₁₁NO₂ Molecular Weight: 165,19 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06804561 CAS-Nummer: 885278-12-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...