1,6-Naphthyridin-2(1H)-one 97%

Lieferant: Thermo Scientific

H50121.03
H50121.03EA 218 EUR
H50121.03
1,6-Naphthyridin-2(1H)-one 97%
1,6-Naphthyridin-2(1H)-one

Purity: 97% CAS No.: 23616-29-7 Molecular Formula: C8H6N2O Formula Weight: 146.15 MDL No.: MFCD07781179

Formel: C₈H₆N₂O
Molecular Weight: 146,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07781179
CAS-Nummer: 23616-29-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR