6-Pyrrolidin-1-ylnicotinaldehyde 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 6-(1-Pyrrolidinyl)nicotinaldehyde

H50126.03
H50126.03EA 162 EUR
H50126.03
6-Pyrrolidin-1-ylnicotinaldehyde 98%
6-Pyrrolidin-1-ylnicotinaldehyde

Purity: 98% CAS No.: 261715-39-3 Molecular Formula: C10H12N2O Formula Weight: 176.22 MDL No.: MFCD06200881

Formel: C₁₀H₁₂N₂O
Molecular Weight: 176,22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06200881
CAS-Nummer: 261715-39-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR