2-Cyclohexyloxypyridine-5-carboxaldehyde 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 6-(Cyclohexyloxy)-3-pyridinecarboxaldehyde

H50129.03
H50129.03EA 72 EUR
H50129.03
2-Cyclohexyloxypyridine-5-carboxaldehyde 97%
2-Cyclohexyloxypyridine-5-carboxaldehyde

Purity: 97% CAS No.: 916792-16-0 Molecular Formula: C12H15NO2 Formula Weight: 205.26 MDL No.: MFCD08060944

Formel: C₁₂H₁₅NO₂
Molecular Weight: 205,26 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08060944
CAS-Nummer: 916792-16-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR