4-Chlor-8-methylchinolin
Lieferant: Thermo Scientific
|
Achtung
|
H50201.03
H50201.03EA
145
EUR
H50201.03
4-Chlor-8-methylchinolin
4-Chloro-8-methylquinoline
CAS No.: 18436-73-2
Molecular Formula: C10H8ClN
Formula Weight: 177.63
MDL No.: MFCD00272330
Formel:
C₁₀H₈ClN Molecular Weight: 177,63 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00272330 CAS-Nummer: 18436-73-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...