3-(4-Chlorphenyl)propionsäure ≥94%

Lieferant: Thermo Scientific
Achtung

Synonyme: 3-(4-Chlorphenyl)propansäure

H50204.03
H50204.03EA 39.9 EUR
H50204.03
3-(4-Chlorphenyl)propionsäure ≥94%
3-(4-Chlorphenyl)propionsäure

Purity: 94% CAS No.: 2019-34-3 Molecular Formula: C9H9ClO2 Formula Weight: 184.62 Melting Point: 118-120° MDL No.: MFCD00016555

Formel: C₉H₉ClO₂
Molecular Weight: 184,62 g/mol
Schmelzpunkt: 118…120 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00016555
CAS-Nummer: 2019-34-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR