Boc-Cys(Et)-OH
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
(R)-2-((tert-Butoxycarbonyl)amino)-3-(ethylthio)propanoic acid
H50216.03
H50216.06
H50216.03EA
86
EUR
H50216.03
H50216.06
Boc-Cys(Et)-OH
Boc-Cys(Et)-OH
CAS No.: 16947-82-3
Molecular Formula: C10H19NO4S
Formula Weight: 249.33
MDL No.: MFCD00076919
Formel:
C₁₀H₁₉NO₄S Molecular Weight: 249.325 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00076919 CAS-Nummer: 16947-82-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...