2-Chlor-N,N-dimethylacetamid ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

H50229.03 H50229.MD H50229.06
H50229.03EA 207 EUR
H50229.03 H50229.MD H50229.06
2-Chlor-N,N-dimethylacetamid ≥97%
2-Chlor-N,N-dimethylacetamid

Purity: 97% CAS No.: 2675-89-0 Molecular Formula: C4H8ClNO Formula Weight: 121.57 Refractive Index: 1.4790 Boiling Point: 98-100°/11mm MDL No.: MFCD00039360

Formel: C₄H₈ClNO
Molecular Weight: 121.57 g/mol
Siedepunkt: 98…100 °C (11 mmHg)
Dichte: 1.182
Lagertemperatur: Raumtemperatur
MDL: MFCD00039360
CAS-Nummer: 2675-89-0
EINECS: 220-224-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR