4-(Methylsulfonyl)benzoesäure ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H50230.03
H50230.03EA
30.2
EUR
H50230.03
4-(Methylsulfonyl)benzoesäure ≥98%
4-(Methylsulphonyl)benzoic acid
Purity: 98%
CAS No.: 4052-30-6
Molecular Formula: C8H8O4S
Formula Weight: 200.21
Melting Point: 270-275°
MDL No.: MFCD00007564
Formel:
C₈H₈O₄S Schmelzpunkt: 270…275 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007564 CAS-Nummer: 4052-30-6 EINECS: 223-765-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...