3-Brom-2-fluorbenzoesäure ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

H50235.06
H50235.06EA 217 EUR
H50235.06
3-Brom-2-fluorbenzoesäure ≥97%
3-Bromo-2-fluorobenzoic acid

Purity: 97% CAS No.: 161957-56-8 Molecular Formula: C7H4BrFO2 Formula Weight: 219.01 Melting Point: 168-170° MDL No.: MFCD00665763

Formel: C₇H₄BrFO₂
Schmelzpunkt: 168…170 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00665763
CAS-Nummer: 161957-56-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR