1-Brom-2-(brommethyl)naphthalin ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

H50240.03
H50240.03EA 120 EUR
H50240.03
1-Brom-2-(brommethyl)naphthalin ≥98%
1-Bromo-2-(bromomethyl)naphthalene

Purity: 98% CAS No.: 37763-43-2 Molecular Formula: C11H8Br2 Formula Weight: 299.99 Melting Point: 108° UN No.: 3261 MDL No.: MFCD00046369

Formel: C₁₁H₈Br₂
Molecular Weight: 299.99 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00046369
CAS-Nummer: 37763-43-2
UN: 3261
ADR: 8 III

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR