(R)-(-)-2-Benzylamino-1-butanol 99%

Lieferant: Thermo Fisher Scientific
Achtung

H50328.03
H50328.03EA 116 EUR
H50328.03
(R)-(-)-2-Benzylamino-1-butanol 99%
(R)-(-)-2-Benzylamino-1-butanol

Purity: 99% CAS No.: 6257-49-4 Molecular Formula: C11H17NO Formula Weight: 179.26 MDL No.: MFCD00085354

Formel: C₁₁H₁₇NO
Molecular Weight: 179.26 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00085354
CAS-Nummer: 6257-49-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR