5-(4-Bromphenyl)isoxazol ≥98%

Lieferant: Thermo Fisher Scientific

H50342.06
H50342.06EA 166 EUR
H50342.06
5-(4-Bromphenyl)isoxazol ≥98%
5-(4-Bromophenyl)isoxazole

Purity: 98% CAS No.: 7064-31-5 Molecular Formula: C9H6BrNO Formula Weight: 224.06 Melting Point: 115-118° MDL No.: MFCD02183532

Formel: C₉H₆BrNO
Molecular Weight: 224,06 g/mol
Schmelzpunkt: 115…118 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02183532
CAS-Nummer: 7064-31-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR