5-Chlorosulphonyl-2-hydroxybenzoic acid 97%
Lieferant: Thermo Scientific
|
Gefahr
|
H50374.06
H50374.06EA
397
EUR
H50374.06
5-Chlorosulphonyl-2-hydroxybenzoic acid 97%
5-Chlorosulphonyl-2-hydroxybenzoic acid
Purity: 97%
CAS No.: 17243-13-9
Molecular Formula: C7H5ClO5S
Formula Weight: 236.63
Melting Point: 171-174°
UN No.: UN2923
MDL No.: MFCD00052974
Formel:
C₇H₅ClO₅S Molecular Weight: 236,63 g/mol Schmelzpunkt: 171…174 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00052974 CAS-Nummer: 17243-13-9 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...