1,2-Benzoldisulfonylchlorid
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
H50375.03
H50375.06
H50375.03EA
122
EUR
H50375.03
H50375.06
1,2-Benzoldisulfonylchlorid
1,2-Benzoldisulfonylchlorid
CAS No.: 6461-76-3
Molecular Formula: C6H4Cl2O4S2
Formula Weight: 275.13
Melting Point: 140-146°
UN No.: 3261
MDL No.: MFCD00059836
Formel:
C₆H₄Cl₂O₄S₂ Schmelzpunkt: 140…146 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00059836 CAS-Nummer: 6461-76-3 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...