3,5-Dichlor-4-hydroxybenzonitril ≥97%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Chloroxynil
H50432.03
H50432.03EA
120
EUR
H50432.03
3,5-Dichlor-4-hydroxybenzonitril ≥97%
3,5-Dichlor-4-hydroxybenzonitril
Purity: 97+%
CAS No.: 1891-95-8
Molecular Formula: C7H3Cl2NO
Formula Weight: 188.01
Melting Point: 143-144°
UN No.: 3439
MDL No.: MFCD00002177
Formel:
C₇H₃Cl₂NO Molecular Weight: 188,01 g/mol Schmelzpunkt: 143…144 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002177 CAS-Nummer: 1891-95-8 EINECS: 217-572-4 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...