8-hydroxyquinoline-2-carbonitrile 98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 8-Hydroxyquinoline-2-carbonitrile

H50433.03 H50433.06
H50433.03EA 69.8 EUR
H50433.03 H50433.06
8-hydroxyquinoline-2-carbonitrile 98%
8-hydroxyquinoline-2-carbonitrile

Purity: 98% CAS No.: 6759-78-0 Molecular Formula: C10H6N2O Formula Weight: 170.17 Melting Point: 133-136° UN No.: UN3439 MDL No.: MFCD00216717

Formel: C₁₀H₆N₂O
Molecular Weight: 170.171 g/mol
Schmelzpunkt: 133…136 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00216717
CAS-Nummer: 6759-78-0
ADR: 6.1 III

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR