3-(3-Bromophenyl)-1H-pyrazole 98%

Lieferant: Thermo Scientific

Synonyme: 3-(3-Bromophenyl)-1H-pyrazole

H50438.06
H50438.06EA 262 EUR
H50438.06
3-(3-Bromophenyl)-1H-pyrazole 98%
3-(3-Bromophenyl)-1H-pyrazole

Purity: 98% CAS No.: 149739-65-1 Molecular Formula: C9H7BrN2 Formula Weight: 223.07 MDL No.: MFCD01940433

Formel: C₉H₇BrN₂
Molecular Weight: 223,07 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01940433
CAS-Nummer: 149739-65-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR