Ethyl-4-chlor-8-methoxychinolin-3-carboxylat
Lieferant: Thermo Scientific
Synonyme:
Ethyl 4-chloro-8-methoxyquinoline-3-carboxylate
H50509.03
H50509.03EA
190
EUR
H50509.03
Ethyl-4-chlor-8-methoxychinolin-3-carboxylat
Ethyl-4-chlor-8-methoxychinolin-3-carboxylat
CAS No.: 27568-05-4
Molecular Formula: C13H12ClNO3
Formula Weight: 265.70
MDL No.: MFCD00173395
Formel:
C₁₃H₁₂ClNO₃ Molecular Weight: 265,70 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00173395 CAS-Nummer: 27568-05-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...