4-Chloro-5-phenylthieno[2,3-d]pyrimidine 98%

Lieferant: Thermo Scientific
Achtung

H50537.03
H50537.03EA 138 EUR
H50537.03
4-Chloro-5-phenylthieno[2,3-d]pyrimidine 98%
4-Chloro-5-phenylthieno[2,3-d]pyrimidine

Purity: 98% CAS No.: 182198-35-2 Molecular Formula: C12H7ClN2S Formula Weight: 246.72 Melting Point: 125-129° MDL No.: MFCD00218029

Formel: C₁₂H₇ClN₂S
Molecular Weight: 246,72 g/mol
Schmelzpunkt: 125…129 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00218029
CAS-Nummer: 182198-35-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR