2-Brom-5-methoxybenzonitril ≥98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 2-Brom-5-methoxybenzoesäurenitril

H50608.MD
H50608.MDEA 77.1 EUR
H50608.MD
2-Brom-5-methoxybenzonitril ≥98%
2-Brom-5-methoxybenzonitril

Purity: 98% CAS No.: 138642-47-4 Molecular Formula: C8H6BrNO Formula Weight: 212.05 UN No.: 3439 MDL No.: MFCD01861260

Formel: C₈H₆BrNO
Molecular Weight: 212,05 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01861260
CAS-Nummer: 138642-47-4
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR