Ethyl-(2-bromphenyl)acetat ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H50630.03
H50630.MD
H50630.03EA
114
EUR
H50630.03
H50630.MD
Ethyl-(2-bromphenyl)acetat ≥98%
Ethyl-(2-bromphenyl)acetat
Purity: 98%
CAS No.: 2178-24-7
Molecular Formula: C10H11BrO2
Formula Weight: 243.10
Melting Point: 35-38°
MDL No.: MFCD00051921
Formel:
C₁₀H₁₁BrO₂ Schmelzpunkt: 35…38 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00051921 CAS-Nummer: 2178-24-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...