3-Methyl-5-phenyl-1H-pyrazol

Lieferant: Thermo Scientific

Synonyme: 5-Methyl-3-phenyl-1H-pyrazole

H50650.03
H50650.03EA 131 EUR
H50650.03
3-Methyl-5-phenyl-1H-pyrazol
3-Methyl-5-phenyl-1H-pyrazole

CAS No.: 3347-62-4 Molecular Formula: C10H10N2 Formula Weight: 158.20 Melting Point: 124-127° MDL No.: MFCD00022385

Formel: C₁₀H₁₀N₂
Molecular Weight: 158,2 g/mol
Schmelzpunkt: 124…127 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00524752
CAS-Nummer: 3347-62-4
EINECS: 222-099-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR