3-(2-Pyridyl)-5-(4-pyridyl)-1,2,4-triazole 99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H50665.03
H50665.03EA
124
EUR
H50665.03
3-(2-Pyridyl)-5-(4-pyridyl)-1,2,4-triazole 99%
3-(2-Pyridyl)-5-(4-pyridyl)-1,2,4-triazole
Purity: 99%
CAS No.: 36770-50-0
Molecular Formula: C12H9N5
Formula Weight: 223.24
MDL No.: MFCD00107797
Formel:
C₁₂H₉N₅ Molecular Weight: 223.24 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00107797 CAS-Nummer: 36770-50-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...