5-Methyl-3-isoxazolcarbonylchlorid ≥98%

Lieferant: Thermo Scientific
Gefahr

H50693.03
H50693.03EA 89.2 EUR
H50693.03
5-Methyl-3-isoxazolcarbonylchlorid ≥98%
5-Methyl-3-isoxazolcarbonylchlorid

Purity: 98% CAS No.: 39499-34-8 Molecular Formula: C5H4ClNO2 Formula Weight: 145.55 UN No.: 3265 MDL No.: MFCD00085127

Formel: C₅H₄ClNO₂
Molecular Weight: 145,55 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00085127
CAS-Nummer: 39499-34-8
EINECS: 254-475-6
UN: 3265
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR