3-(2-Methylphenoxy)pyrrolidine 95%

Lieferant: Thermo Fisher Scientific
Achtung

H50784.03 H50784.MD
H50784.03EA 135 EUR
H50784.03 H50784.MD
3-(2-Methylphenoxy)pyrrolidine 95%
3-(2-Methylphenoxy)pyrrolidine

Purity: 95% CAS No.: 46196-54-7 Molecular Formula: C11H15NO Formula Weight: 177.25 MDL No.: MFCD08060984

Formel: C₁₁H₁₅NO
Lagertemperatur: Raumtemperatur
MDL: MFCD08060984
CAS-Nummer: 46196-54-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR