4-[4-(4-Chlorophenyl)-1H-pyrazol-1-yl]piperidine

Lieferant: Thermo Fisher Scientific
Achtung

H50804.03
H50804.03EA 192 EUR
H50804.03
4-[4-(4-Chlorophenyl)-1H-pyrazol-1-yl]piperidine
4-[4-(4-Chlorophenyl)-1H-pyrazol-1-yl]piperidine

CAS No.: 902836-38-8 Molecular Formula: C14H16ClN3 Formula Weight: 261.76 MDL No.: MFCD08060997

Formel: C₁₄H₁₆ClN₃
Lagertemperatur: Raumtemperatur
MDL: MFCD08060997
CAS-Nummer: 902836-38-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR