1-(3-Chlor-2-pyridinyl)piperazin ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

H50859.03 H50859.MD
H50859.03EA 227 EUR
H50859.03 H50859.MD
1-(3-Chlor-2-pyridinyl)piperazin ≥98%
1-(3-Chlor-2-pyridinyl)piperazin

Purity: 98% CAS No.: 87394-55-6 Molecular Formula: C9H12ClN3 Formula Weight: 197.67 MDL No.: MFCD08061034

Formel: C₉H₁₂ClN₃
Lagertemperatur: Raumtemperatur
MDL: MFCD08061034
CAS-Nummer: 87394-55-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR