Benzo[d]isoxazol-3-ol 96%

Lieferant: Thermo Fisher Scientific
Achtung

H50899.03 H50899.MD
H50899.03EA 193 EUR
H50899.03 H50899.MD
Benzo[d]isoxazol-3-ol 96%
Benzo[d]isoxazol-3-ol

Purity: 96% CAS No.: 21725-69-9 Molecular Formula: C7H5NO2 Formula Weight: 135.12 Odour: Odourless Melting Point: 137-140° MDL No.: MFCD00125030

Formel: C₇H₅NO₂
Schmelzpunkt: 137…140 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00125030
CAS-Nummer: 21725-69-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR