1-Pyridin-2-yl[1,4]diazepane ≥98%

Lieferant: Thermo Scientific

H50946.MD
H50946.MDEA 0 EUR
H50946.MD
1-Pyridin-2-yl[1,4]diazepane ≥98%
1-Pyridin-2-yl[1,4]diazepane

Purity: 98% CAS No.: 287114-32-3 Molecular Formula: C10H15N3 Formula Weight: 177.25 Storage Temperature: 2°C to 8°C Boiling Point: 104°/0.4mm UN No.: UN2735 MDL No.: MFCD06409204

Formel: C₁₀H₁₅N₃
MDL: MFCD06409204
CAS-Nummer: 287114-32-3
ADR: ,

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR