4-(3-Methylphenoxy)piperidin ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-(m-Tolyloxy)piperidine

H50950.03
H50950.03EA 133 EUR
H50950.03
4-(3-Methylphenoxy)piperidin ≥97%
4-(3-Methylphenoxy)piperidin

Purity: 97% CAS No.: 63843-46-9 Molecular Formula: C12H17NO Formula Weight: 191.28 Melting Point: 190-191° MDL No.: MFCD06248345

Formel: C₁₂H₁₇NO
Molecular Weight: 191,28 g/mol
Schmelzpunkt: 190…191 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06248345
CAS-Nummer: 63843-46-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR