4-(3-Chlorphenoxy)piperidin ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
H50956.03
H50956.03EA
227
EUR
H50956.03
4-(3-Chlorphenoxy)piperidin ≥98%
4-(3-Chlorphenoxy)piperidin
Purity: 98%
CAS No.: 97840-40-9
Molecular Formula: C11H14ClNO
Formula Weight: 211.69
MDL No.: MFCD08061087
Formel:
C₁₁H₁₄ClNO Molecular Weight: 211,69 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08061087 CAS-Nummer: 97840-40-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...