4-(4-Piperidinyloxy)benzonitril ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

H50958.03 H50958.MD
H50958.03EA 245 EUR
H50958.03 H50958.MD
4-(4-Piperidinyloxy)benzonitril ≥98%

Purity: 98% CAS No.: 224178-67-0 Molecular Formula: C12H14N2O Formula Weight: 202.26 UN No.: 3439 MDL No.: MFCD07368391

Formel: C₁₂H₁₄N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD07368391
CAS-Nummer: 224178-67-0
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR