2-(4-Chlorophenyl)-2-methylmorpholine 99%

Lieferant: Thermo Fisher Scientific
Achtung

H51010.MD
H51010.MDEA 40.4 EUR
H51010.MD
2-(4-Chlorophenyl)-2-methylmorpholine 99%
2-(4-Chlorophenyl)-2-methylmorpholine

Purity: 99% CAS No.: 109461-44-1 Molecular Formula: C11H14ClNO Formula Weight: 211.69 MDL No.: MFCD08061115

Formel: C₁₁H₁₄ClNO
Lagertemperatur: Raumtemperatur
MDL: MFCD08061115
CAS-Nummer: 109461-44-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR