(2S,4R)-1-(tert-Butoxycarbonyl)-4-hydroxy-2-(hydroxymethyl)pyrrolidine 96%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
tert-Butyl (2S,4R)-4-hydroxy-2-hydroxymethylpyrrolidine-1-carboxylate, N-Boc-trans-4-hydroxy-L-prolinol, (2S,4R)-1-Boc-4-hydroxy-2-(hydroxymethyl)pyrrolidine, Boc-Hyp-OL
H51050.06
H51050.06EA
248
EUR
H51050.06
(2S,4R)-1-(tert-Butoxycarbonyl)-4-hydroxy-2-(hydroxymethyl)pyrrolidine 96%
(2S,4R)-1-(tert-Butoxycarbonyl)-4-hydroxy-2-(hydroxymethyl)pyrrolidine
Purity: 96%
CAS No.: 61478-26-0
Molecular Formula: C10H19NO4
Formula Weight: 217.26
MDL No.: MFCD02094386
Formel:
C₁₀H₁₉NO₄ Molecular Weight: 217,27 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02094386 CAS-Nummer: 61478-26-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...