(R)-(+)-4-(Diphenylmethyl)-2-oxazolidinone 97%

Lieferant: Thermo Scientific

Synonyme: (R)-4-Benzhydryloxazolidin-2-one

H51074.03
H51074.03EA 261 EUR
H51074.03
(R)-(+)-4-(Diphenylmethyl)-2-oxazolidinone 97%
(R)-(+)-4-(Diphenylmethyl)-2-oxazolidinone

Purity: 98% CAS No.: 173604-33-6 Molecular Formula: C16H15NO2 Formula Weight: 253.30 MDL No.: MFCD01863565

Formel: C₁₆H₁₅NO₂
Molecular Weight: 253,3000 g/mol
MDL: MFCD01863565
CAS-Nummer: 173604-33-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR