(R)-(+)-2-Amino-1,1-diphenyl-1-propanol 99%

Lieferant: Thermo Scientific
Gefahr

Synonyme: (R)-2-Amino-1,1-diphenylpropan-1-ol

H51083.03
H51083.03EA 144 EUR
H51083.03
(R)-(+)-2-Amino-1,1-diphenyl-1-propanol 99%
(R)-(+)-2-Amino-1,1-diphenyl-1-propanol

Purity: 99% CAS No.: 78603-93-7 Molecular Formula: C15H17NO Formula Weight: 227.30 Melting Point: 102-105° UN No.: UN3259 MDL No.: MFCD02677967

Formel: C₁₅H₁₇NO
Molecular Weight: 227,30 g/mol
Schmelzpunkt: 102…105 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02677967
CAS-Nummer: 78603-93-7
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR