5-Chinolinol ≥99%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
5-Hydroxyquinoline, Quinolin-5-ol
H51096.MD
H51096.MDEA
25.6
EUR
H51096.MD
5-Chinolinol ≥99%
5-Chinolinol
Purity: 99%
CAS No.: 578-67-6
Molecular Formula: C9H7NO
Formula Weight: 145.16
MDL No.: MFCD00006792
Formel:
C₉H₇NO Molecular Weight: 145,16 g/mol Siedepunkt: 313 °C (1013 hPa) Schmelzpunkt: 223…226 °C Dichte: 1,57 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006792 CAS-Nummer: 578-67-6 EINECS: 209-428-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...